Learn More
Invitrogen™ SuperNuclease Recombinant Rabbit Monoclonal Antibody (026), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
271.00 CHF - 544.00 CHF
Spezifikation
| Antigen | SuperNuclease |
|---|---|
| Klon | 026 |
| Konzentration | 1 mg/mL |
| Anwendungen | ELISA |
| Klassifikation | Recombinant Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
17799793
|
Invitrogen™
MA543451 |
50 μL |
271.00 CHF
50 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
17799643
![]() |
Invitrogen™
MA543096 |
100 μL |
544.00 CHF
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Specificity: SuperNuclease (Recombinant Serratia marcescens Endonuclease)
SuperNuclease is a genetically-engineered endonuclease. It is a nonspecific nuclease with high activity, capable of completely digesting RNA and DNA (single-stranded, double-stranded, linear, circular and supercoiled forms, that no fewer than five phosphate residues) into 5'-monophosphate-terminated oligonucleotides of 3-5 bases in length. SuperNuclease can be removed by various purification methods.Spezifikation
| SuperNuclease | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| SuperNuclease (Recombinant Serratia marcescens Endonuclease). | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles |
| 026 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Bacteria | |
| DNA endonuclease; RNA endonuclease | |
| IgG | |
| Protein A | |
| Antibody |