missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SuperNuclease Recombinant Rabbit Monoclonal Antibody (026), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA543096
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Specificity: SuperNuclease (Recombinant Serratia marcescens Endonuclease)
SuperNuclease is a genetically-engineered endonuclease. It is a nonspecific nuclease with high activity, capable of completely digesting RNA and DNA (single-stranded, double-stranded, linear, circular and supercoiled forms, that no fewer than five phosphate residues) into 5'-monophosphate-terminated oligonucleotides of 3-5 bases in length. SuperNuclease can be removed by various purification methods.
Spezifikation
| SuperNuclease | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| ELISA | |
| 026 | |
| Unconjugated | |
| DNA endonuclease; RNA endonuclease | |
| SuperNuclease (Recombinant Serratia marcescens Endonuclease). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur