missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21624-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Striatin is found in all mammalian cells assayed and may be involved in vesicular transport, dendrite growth, and cellular signaling. It binds Protein phosphatase 2A (PP2A) A and C subunits and calmodulin and appears to modulate PP2A activity. It also binds the human homolog of the yeast protein, Mob1, which has been implicated in mitotic progression in yeast. It is a member of a family of proteins that also includes SG2NA and Zinedin. This antibody does not cross-react with SG2NA.Spezifikation
STRN | |
Polyclonal | |
Unconjugated | |
STRN | |
SG2NA, Striatin, STRN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
268980, 29149, 6801 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43815, O55106, P70483 | |
Strn | |
STRN Fusion Protein Ag15984 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |