missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | STRN |
---|---|
Konzentration | 0.2 mg/mL |
Anwendungen | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16851225
|
Proteintech
21624-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16841225
|
Proteintech
21624-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Striatin is found in all mammalian cells assayed and may be involved in vesicular transport, dendrite growth, and cellular signaling. It binds Protein phosphatase 2A (PP2A) A and C subunits and calmodulin and appears to modulate PP2A activity. It also binds the human homolog of the yeast protein, Mob1, which has been implicated in mitotic progression in yeast. It is a member of a family of proteins that also includes SG2NA and Zinedin. This antibody does not cross-react with SG2NA.Spezifikation
STRN | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O43815, O55106, P70483 | |
268980, 29149, 6801 | |
STRN Fusion Protein Ag15984 | |
Primary | |
-20°C | |
STRN |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
SG2NA, Striatin, STRN | |
Strn | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |