missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STATH Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19724-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
STATH is a salivary protein that stabilizes saliva supersaturated with calcium salts by inhibiting the precipitation of calcium phosphate salts. It also modulates hydroxyapatite crystal formation on the tooth surface.Spezifikation
STATH | |
Polyclonal | |
Unconjugated | |
STATH | |
STATH, statherin, STR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6779 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02808 | |
STATH | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |