missing translation for 'onlineSavingsMsg'
Learn More

STAT2 Mouse anti-Human, Clone: 1G12C4, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66485-1-IG-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838405

  • 351.68 CHF / 150 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

CD95 (Fas, APO-1), a 46 kDa transmembrane glycoprotein, is a cell death receptor of the TNFR superfamily. Stimulation of CD95 results in aggregation of its intracellular death domains, formation of the death-inducing signaling complex (DISC) and activation of caspases. In type I cells caspase 3 is activated by high amounts of caspase 8 generated at the DISC, in type II cells low concentration of caspase 8 activates pathway leading to the release of cytochrome c from mitochondria and activation of caspase 3 by cytochom c. Besides its roles in induction of apoptosis, Fas also triggers pro-inflammatory cytokine responses.
Spezifikation

Spezifikation

STAT2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P52630
STAT2
STAT2 Fusion Protein Ag10168
150 μL
Primary
Human
Liquid
Western Blot
1G12C4
Unconjugated
STAT2
ISGF 3, P113, STAT113, STAT2
Mouse
Protein G
RUO
6773
Antibody
IgG1
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote