missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT2 Mouse anti-Human, Clone: 1G12C4, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66485-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
CD95 (Fas, APO-1), a 46 kDa transmembrane glycoprotein, is a cell death receptor of the TNFR superfamily. Stimulation of CD95 results in aggregation of its intracellular death domains, formation of the death-inducing signaling complex (DISC) and activation of caspases. In type I cells caspase 3 is activated by high amounts of caspase 8 generated at the DISC, in type II cells low concentration of caspase 8 activates pathway leading to the release of cytochrome c from mitochondria and activation of caspase 3 by cytochom c. Besides its roles in induction of apoptosis, Fas also triggers pro-inflammatory cytokine responses.Spezifikation
STAT2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P52630 | |
STAT2 | |
STAT2 Fusion Protein Ag10168 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
1G12C4 | |
Unconjugated | |
STAT2 | |
ISGF 3, P113, STAT113, STAT2 | |
Mouse | |
Protein G | |
RUO | |
6773 | |
-20°C | |
Liquid |