missing translation for 'onlineSavingsMsg'
Learn More

SSEA-1 Mouse, PE, Clone: MC480, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 560142

Additional Details : Gewicht : 0.02200kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15819968

  • 575.00 CHF / 1 Stück
Verfügbar ab: 17-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The MC480 monoclonal antibody reacts with Stage-Specific Embryonic Antigen-1 (SSEA-1), which is a terminal carbohydrate epitope (3-fucosyl-N-acetyllactosamine or 3-FAL) on glycoproteins and lactose series glycolipids. SSEA-1 is related to Lewis blood group antigens and is found in a variety of embryonic and adult tissues and cancers. As its name implies, the expression of SSEA-1 is stage-specific and can be used to characterize embryonic cells and monitor their differentiation. However, its expression pattern differs in the human and mouse. In the human, SSEA-1 is not found on embryonic stem (ES) cells, embryonic inner cell mass (ICM), or teratocarcinoma (embryonal carcinoma or EC) cells. As human EC and ES cells undergo differentiation, SSEA-1 expression is upregulated. In the adult, the same epitope is expressed as CD15 on granulocytes and monocytes, but not lymphocytes or dendritic cells. In the mouse, SSEA-1 is found on EC, ES, and primordial germ cells, 8-cell to blastocyst embryos, ICM, and on subpopulations of cells in the adult central nervous system, including stem cells. In contrast to the human, SSEA-1 expression is reduced as mouse EC and ES cells undergo differentiation.

Host Species: Mouse
Clone: MC480
Isotype: IgM, κ
Species Reactivity: Mouse
Immunogen: Mouse Teratocarcinoma Cell Line

Flow Cytometry

Spezifikation

Spezifikation

SSEA-1
Monoclonal
PE
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Mouse Teratocarcinoma Cell Line
100 Tests
Cell Biology
Human, Mouse
IgM κ
Flow Cytometry
MC480
3-FAL, X-hapten, LeX antigen, CD15
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.