missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRP54 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11729-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Binds to the signal sequence of presecretory protein when they emerge from the ribosomes and transfers them to TRAM (translocating chain-associating membrane protein).Spezifikation
SRP54 | |
Polyclonal | |
Unconjugated | |
SRP54 | |
SRP54 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116650, 24067, 6729 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P14576, P61011, Q6AYB5 | |
SRP54 | |
SRP54 Fusion Protein Ag2327 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |