missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SRC, His Tag Recombinant Protein
SRC Recombinant Human Protein
Marke: Invitrogen™ PR4336E
Additional Details : Gewicht : 0.01000kg
Beschreibung
SRC is involved in the regulation of embryonic development and cell growth.
Spezifikation
NP_005408 | |
62.3 kDa | |
SRC | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Full Length, Recombinant, Cytoplasmic | |
Cytoplasmic Tyrosine Kinases |
Kinase Assay | |
100 μg | |
SRC Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 100 mM NaCl, 0.05 mM EDTA, 0.05% NP-40, 1 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.