missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SRC, His Tag Recombinant Protein
SRC Recombinant Human Protein
Marke: Invitrogen™ P3044
Additional Details : Gewicht : 3.36000kg
Beschreibung
SRC is a cytoplasmic tyrosine kinase and proto-oncogene involved in the regulation of embryonic development and cell growth.Spezifikation
NP_005408 | |
6714 | |
10μg | |
SRC Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 100 mM NaCl, 0.05 mM EDTA, 0.05% NP-40, 1 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
10 μg | |
Dry Ice | |
C-Terminal | |
v-src sarcoma | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
62.3 kDa | |
SRC | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
SRC | |
Baculovirus | |
Full Length, Recombinant, Cytoplasmic | |
Cytoplasmic Tyrosine Kinases |
For Research Use Only. Not for use in diagnostic procedures.