missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR140 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21399-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
TMEM184B, also known as C22orf5, is a 407 amino acid multi-pass membrane protein and represents a novel gene in the activation of the MAPK signaling pathway. The gene encoding TMEM184B maps to human chromosome 22; mutations in several of the genes in chromosome 22 are involved in the development of autism, schizophrenia, Phelan-McDermid syndrome and Neurofibromatosis type 2, suggesting that TMEM184B may play a role in these syndromes.Spezifikation
SR140 | |
Polyclonal | |
Unconjugated | |
U2SURP | |
KIAA0332, SR140, U2 associated protein SR140, U2 associated SR140 protein, U2SURP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23350, 315903, 67958 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15042, Q6NV83 | |
U2surp | |
SR140 Fusion Protein Ag16093 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |