missing translation for 'onlineSavingsMsg'
Learn More

SR140 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 21399-1-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16841005

  • 381.34 CHF / 150 Mikroliter
Verfügbar ab: 16-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

TMEM184B, also known as C22orf5, is a 407 amino acid multi-pass membrane protein and represents a novel gene in the activation of the MAPK signaling pathway. The gene encoding TMEM184B maps to human chromosome 22; mutations in several of the genes in chromosome 22 are involved in the development of autism, schizophrenia, Phelan-McDermid syndrome and Neurofibromatosis type 2, suggesting that TMEM184B may play a role in these syndromes.
Spezifikation

Spezifikation

SR140
Polyclonal
Unconjugated
U2SURP
KIAA0332, SR140, U2 associated protein SR140, U2 associated SR140 protein, U2SURP
Rabbit
Antigen Affinity Chromatography
RUO
23350, 315903, 67958
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.44 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15042, Q6NV83
U2surp
SR140 Fusion Protein Ag16093
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote