missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12020-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody can recognize all 3 isoforms of SPATA7.
The function of the SPATA7 protein remains unknown.Spezifikation
SPATA7 | |
Polyclonal | |
Unconjugated | |
SPATA7 | |
HSD 3.1, HSD3, SPATA7, spermatogenesis associated 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104871, 55812 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80VP2, Q9P0W8 | |
SPATA7 | |
SPATA7 Fusion Protein Ag2649 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |