missing translation for 'onlineSavingsMsg'
Learn More

SORCS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 18839-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838694

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 22-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This antibody was generated against the C-terminal region of human SorCS1 and recognizes various forms of SorCS1. Multiple bands around 100-130 kDa as well as 60-80 kDa have been detected in Y79 cells by western blot assay.

SORCS1 is one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein.
Spezifikation

Spezifikation

SORCS1
Polyclonal
Unconjugated
SORCS1
hSorCS, SORCS1
Rabbit
Antigen Affinity Chromatography
RUO
114815, 58178
-20°C
Liquid
Western Blot, Flow Cytometry
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WY21, Q9JLC4
Sorcs1
Peptide
20 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote