Learn More
SORCS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18839-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody was generated against the C-terminal region of human SorCS1 and recognizes various forms of SorCS1. Multiple bands around 100-130 kDa as well as 60-80 kDa have been detected in Y79 cells by western blot assay.
SORCS1 is one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein.Spezifikation
SORCS1 | |
Polyclonal | |
Unconjugated | |
SORCS1 | |
hSorCS, SORCS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114815, 58178 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WY21, Q9JLC4 | |
Sorcs1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |