Learn More
SOD1 Mouse, anti-Human, CoraLite 594, Clone: 2F10G1, Proteintech
Mouse Monoclonal Antibody
136.48 CHF - 423.57 CHF
Spezifikation
| Antigen | SOD1 |
|---|---|
| Klon | 2F10G1 |
| Konzentration | 1000 μg/mL |
| Anwendungen | Flow Cytometry (Intracellular Staining) |
| Konjugat | CoraLite 594 |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18059795
|
Proteintech
CL594-67480-20UL |
20 μL |
136.48 CHF
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18016185
|
Proteintech
CL594-67480-100UL |
100 μL |
423.57 CHF
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
The SOD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SOD1. This antibody recognizes human, mouse, rat, pig antigen. The SOD1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™594.
Spezifikation
| SOD1 | |
| 1000 μg/mL | |
| CoraLite 594 | |
| Mouse | |
| 6647 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2F10G1 | |
| Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat, Pig | |
| ALS1, homodimer, IPOA, SOD, SOD1, Superoxide dismutase [Cu Zn] | |
| Primary |