Learn More
SOD1 Mouse, anti-Human, CoraLite 594, Clone: 2F10G1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL594-67480-20UL
Beschreibung
The SOD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SOD1. This antibody recognizes human, mouse, rat, pig antigen. The SOD1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™594.
Spezifikation
| SOD1 | |
| 2F10G1 | |
| CoraLite 594 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 155 kDa | |
| IgG2a |
| Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| ALS1, homodimer, IPOA, SOD, SOD1, Superoxide dismutase [Cu Zn] | |
| 20 μL | |
| 6647 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |