Learn More
Novus Biologicals™ SNAP45 Overexpression Lysate
Artikelnummer. 18043649 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
0,1 mg
Packungsgröße:
0.10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18043649 0,1 mg 0.10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18043649 Lieferant Novus Biologicals™ Lieferanten-Nr. NBP210866
 Eingeschränkter Artikel
Verfügbar ab: 12-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
The SNAP45 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifica
Zelllinie The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_374. The protein contains a C-terminal DDK tag.
Zur Verwendung mit (Anwendung) Western Blot
Menge 0,1 mg
Lagerungspuffer RIPA buffer
Reinheit Protein
Testspezifität Homo sapiens SNAP45, mRNA.
Inhalt und Lagerung -80°C. Avoid freeze-thaw cycles.
Protein State Native
Nur für Forschungszwecke
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.