missing translation for 'onlineSavingsMsg'
Learn More

SMAD3 Mouse anti-Human, Mouse, Clone: 1F10B2, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66516-1-IG-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16848465

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 17-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

SMAD3 belongs to the SMAD family of proteins. They are similar to the gene products of the Drosophila gene ′mothers against decapentaplegic′ (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions as a transcriptional modulator activated by transforming growth factor-beta and is thought to play a role in the regulation of carcinogenesis.
Spezifikation

Spezifikation

SMAD3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P84022, Q8BUN5
Smad3
Peptide
20 μL
Primary
Mouse, Human
Antibody
IgG1
Western Blot
1F10B2
Unconjugated
SMAD3
hMAD 3, hSMAD3, HSPC193, HsT17436, JV15 2, MAD homolog 3, Mad3, MADH3, Mothers against DPP homolog 3, SMAD 3, SMAD family member 3, SMAD3
Mouse
Protein G
RUO
17127, 4088
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote