missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC27A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12943-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
SLC27A3 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.SLC27A3 does not exhibit fatty acid transport activity.Spezifikation
SLC27A3 | |
Polyclonal | |
Unconjugated | |
SLC27A3 | |
ACSVL3, FATP 3, FATP3, Fatty acid transport protein 3, SLC27A3, UNQ367/PRO703, VLCS 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11000, 26568, 295219 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88561, Q5K4L6 | |
Slc27a3 | |
SLC27A3 Fusion Protein Ag4015 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |