missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC13A4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20302-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
SLC13A4 gene ontology annotations related to this gene include sodium ion transport; transmembrane transport.Spezifikation
SLC13A4 | |
Polyclonal | |
Unconjugated | |
SLC13A4 | |
NaS2, SLC13A4, SUT 1, SUT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
243755, 26266, 503568 | |
Antibody | |
IgG |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UKG4 | |
Slc13a4 | |
SLC13A4 Fusion Protein Ag14116 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Liquid |