Learn More
Siglec-2/CD22 Antibody, Novus Biologicals™
Artikelnummer. 18472691 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
25 μL
0.1 mL
Packungsgröße:
0.10 Milliliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18472691 25 μL 25 Mikroliter
18221708 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18472691 Lieferant Novus Biologicals Lieferanten-Nr. NBP18704125ul
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
Siglec-2/CD22 Polyclonal specifically detects Siglec-2/CD22 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Siglec-2/CD22
Anwendungen Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Zusammensetzung PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gen-Alias B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14
Gensymbole CD22
Wirtsspezies Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:DLESFILFHNPEYNKNTSKFDGTRLYESTKDGKVPSEQKRVQFLGDKNKNCTLSIHPVHLNDSGQLGLRMESKTEKWMERIHLNVSERPFPPHIQLPPEIQESQEVTLTCLLNFSCYGYPIQLQWLL
Reinigungsverfahren Affinity Purified
Menge 25 μL
Regulatorischer Status RUO
Forschungsgebiet Adaptive Immunity, DNA Repair, Immunology
Primär oder sekundär Primary
Gen-ID (Entrez) 933
Testspezifität Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Zielspezies Human
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG