missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3BGR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21161-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
SH3BGR is a protein coding gene.Spezifikation
SH3BGR | |
Polyclonal | |
Unconjugated | |
SH3BGR | |
21 GARP, 21 glutamic acid rich protein, SH3BGR, SH3BGR protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
50795, 6450 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P55822, Q9WUZ7 | |
Sh3bgr | |
SH3BGR Fusion Protein Ag14739 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |