missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | SH2D1B |
---|---|
Konzentration | 0.37 mg/mL |
Anwendungen | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839233
|
Proteintech
11871-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829233
|
Proteintech
11871-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
SH2D1B plays a role in controlling signal transduction through at least four receptors, CD84, CD150, CD229 and CD244, expressed on the surface of professional antigen-presenting cells.Spezifikation
SH2D1B | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
O14796 | |
117157, 26904, 501869 | |
SH2D1B Fusion Protein Ag2454 | |
Primary | |
-20°C | |
SH2D1B |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EAT 2, EAT2, SH2 domain containing 1B, SH2D1B | |
Sh2d1b, Sh2d1b1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |