missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11871-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
SH2D1B plays a role in controlling signal transduction through at least four receptors, CD84, CD150, CD229 and CD244, expressed on the surface of professional antigen-presenting cells.Spezifikation
SH2D1B | |
Polyclonal | |
Unconjugated | |
SH2D1B | |
EAT 2, EAT2, SH2 domain containing 1B, SH2D1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117157, 26904, 501869 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14796 | |
Sh2d1b, Sh2d1b1 | |
SH2D1B Fusion Protein Ag2454 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |