missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERINC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20265-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody is specific to SERINC1.
SERINC1, also named as KIAA1253, TDE1L and TDE2, belongs to the TDE1 family. SERINC1 enhances the incorporation of serine into phosphatidylserine and sphingolipids.Spezifikation
SERINC1 | |
Polyclonal | |
Unconjugated | |
SERINC1 | |
KIAA1253, SERINC1, serine incorporator 1, TDE1L, TDE2, TMS 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57515 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NRX5 | |
SERINC1 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |