missing translation for 'onlineSavingsMsg'
Learn More

SEPT7 Mouse anti-Human, Mouse, Rat, Clone: 1G12D1, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66542-1-IG-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16848515

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 21-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.
Spezifikation

Spezifikation

SEPT7
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O55131, Q16181, Q9WVC0
SEPT7
SEPT7 Fusion Protein Ag4309
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
1G12D1
Unconjugated
SEPT7
CDC10, CDC10 protein homolog, CDC3, Nbla02942, SEPT7, SEPT7A, septin 7
Mouse
Protein G
RUO
235072, 64551, 989
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote