missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT7 Mouse anti-Human, Mouse, Rat, Clone: 1G12D1, Proteintech
Mouse Monoclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | SEPT7 |
---|---|
Klon | 1G12D1 |
Konzentration | 1 mg/mL |
Anwendungen | Western Blot, Immunohistochemistry (Paraffin) |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16848515
|
Proteintech
66542-1-IG-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16838515
|
Proteintech
66542-1-IG-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.Spezifikation
SEPT7 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CDC10, CDC10 protein homolog, CDC3, Nbla02942, SEPT7, SEPT7A, septin 7 | |
SEPT7 | |
IgG1 | |
Protein G | |
Antibody |
1G12D1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Rat, Mouse, Human | |
O55131, Q16181, Q9WVC0 | |
235072, 64551, 989 | |
SEPT7 Fusion Protein Ag4309 | |
Primary | |
-20°C | |
SEPT7 |