missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT7 Mouse anti-Human, Mouse, Rat, Clone: 1G12D1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66542-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.Spezifikation
SEPT7 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O55131, Q16181, Q9WVC0 | |
SEPT7 | |
SEPT7 Fusion Protein Ag4309 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1G12D1 | |
Unconjugated | |
SEPT7 | |
CDC10, CDC10 protein homolog, CDC3, Nbla02942, SEPT7, SEPT7A, septin 7 | |
Mouse | |
Protein G | |
RUO | |
235072, 64551, 989 | |
-20°C | |
Liquid |