missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCPx Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
131.19 CHF - 351.68 CHF
Spezifikation
Antigen | SCPx |
---|---|
Konzentration | 0.25 mg/mL |
Anwendungen | Immunocytochemistry, Immunofluorescence, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16838914
|
Proteintech
19182-1-AP-20UL |
20 μL |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
16828914
|
Proteintech
19182-1-AP-150UL |
150 μL |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
19182-1-AP antibody recognizes the full-length protein andthe thiolasedomain, as well as the 35 kDa isoform of SCPx.
This gene encodes two proteins: sterol carrier protein X (SCPx) and sterol carrier protein 2 (SCP2), as a result of transcription initiation from 2 independently regulated promoters. The transcript initiated from the proximal promoter encodes the longer SCPx protein, and the transcript initiated from the distal promoter encodes the shorter SCP2 protein, with the 2 proteins sharing a common C-terminus. Evidence suggests that the SCPx protein is a peroxisome-associated thiolase that is involved in the oxidation of branched chain fatty acids, while the SCP2 protein is thought to be an intracellular lipid transfer protein. This gene is highly expressed in organs involved in lipid metabolism, and may play a role in Zellweger syndrome, in which cells are deficient in peroxisomes and have impaired bile acid synthesis. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms. The full-length nature of all transcript variants has not been determined.Spezifikation
SCPx | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P11915, P22307, P32020 | |
20280, 25541, 6342 | |
SCPx Fusion Protein Ag6002 | |
Primary | |
-20°C | |
SCP2 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NLTP, NSL TP, SCP 2, SCP chi, SCP X, SCP2, sterol carrier protein 2, Sterol carrier protein X | |
SCP2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |