missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMD3 Mouse anti-Human, Rat, Clone: 1A11F2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66599-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The function of this protein remains unknown.Spezifikation
SAMD3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N6K7 | |
Samd3 | |
SAMD3 Fusion Protein Ag11319 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Western Blot | |
1A11F2 | |
Unconjugated | |
SAMD3 | |
SAMD3 | |
Mouse | |
Protein G | |
RUO | |
154075, 685940 | |
-20°C | |
Liquid |