missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
The S1P3/EDG-3 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
| Zelllinie | The lysate was created in HEK293T cells, using plasmid ID RC29253 and based on accession number NM_5226. The protein contains a C-terminal DDK tag. |
| Zur Verwendung mit (Anwendung) | Western Blot |
| Menge | 0,1 mg |
| Lagerungspuffer | RIPA buffer |
| Reinheit | Protein |
| Testspezifität | Homo sapiens sphingosine-1-phosphate receptor 3 (S1PR3), mRNA. |
| Inhalt und Lagerung | -80°C. Avoid freeze-thaw cycles. |
| Forschungskategorie | Cell Cycle and Replication, GPCR |
| Protein State | Native |
Nur für Forschungszwecke
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?