missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12055-2-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
RTN3 is a member of reticulons, which are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.Spezifikation
RTN3 | |
Polyclonal | |
Unconjugated | |
RTN3 | |
ASYIP, HAP, NSP like protein 2, NSP like protein II, NSPL2, NSPLII, reticulon 3, RTN3, RTN3 A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10313, 140945, 20168 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95197, Q6RJR6, Q9ES97 | |
RTN3 | |
RTN3 Fusion Protein Ag2685 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |