missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | RTN3 |
---|---|
Konzentration | 0.27 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839543
|
Proteintech
12055-2-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829543
|
Proteintech
12055-2-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
RTN3 is a member of reticulons, which are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.Spezifikation
RTN3 | |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O95197, Q6RJR6, Q9ES97 | |
10313, 140945, 20168 | |
RTN3 Fusion Protein Ag2685 | |
Primary | |
-20°C | |
RTN3 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ASYIP, HAP, NSP like protein 2, NSP like protein II, NSPL2, NSPLII, reticulon 3, RTN3, RTN3 A1 | |
RTN3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |