missing translation for 'onlineSavingsMsg'
Learn More

ROCK1 Mouse anti-Human, Mouse, Rat, Clone: 3G5E12, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66782-1-IG-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16848975

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 14-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The Rho target protein, Rho-associated coiled coil-containing protein kinase (ROCK)-1 has been found to be a new Caspase-3 substrate. ROCK, one of the effectors of the small GTPase Rho, has recently been shown to contribute significantly to myosin light chain (MLC) activation through two pathways: direct phosphorylation of MLC and phosphorylation of MLC phosphatase, leading to its inhibition. The increase in cellular contractility that is necessary for apoptotic membrane blebbing implies sustained augmentation of MLC phosphorylation. ROCK-1 consists of an amino-terminal kinase domain and an inhibitory cysteine/histidine-rich C-terminal domain that is located within a pleckstrin-homology region. They are joined by a variable region that contains the Rho-binding domain. During apoptosis, ROCK-I is cleaved by caspase-3 at a conserved DETD1113/G sequence and its carboxy-terminal inhibitory domain is removed, resulting in deregulated and constitutive kinase activity. The caspase-3-mediated cleavage and activation of ROCK I induces phosphorylation of MLC and membrane blebbing, one of the first events in the execution phase of apoptosis.
Spezifikation

Spezifikation

ROCK1
Monoclonal
0.8 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P70335, Q13464, Q63644
ROCK1
ROCK1 Fusion Protein Ag16317
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG1
Western Blot
3G5E12
Unconjugated
ROCK1
p160 ROCK 1, P160ROCK, PRO0435, ROCK1
Mouse
Protein G
RUO
19877, 6093, 81762
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote