missing translation for 'onlineSavingsMsg'
Learn More

RNF26 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Artikelnummer. 18477247 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18477247 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18477247 Lieferant Novus Biologicals Lieferanten-Nr. NBP281997AF594

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

RNF26 Polyclonal antibody specifically detects RNF26 in Human, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF26
Anwendungen Western Blot, ELISA
Klassifikation Polyclonal
Konjugat Alexa Fluor 594
Zusammensetzung 50mM Sodium Borate
Gen-Alias MGC2642, ring finger protein 26, ring finger protein with leucine zipper
Wirtsspezies Rabbit
Immunogen RNF26 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human RNF26. Amino Acid Squence: RGRERLNEEEPPGGQDPWK
Reinigungsverfahren Peptide affinity purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Zinc Finger
Primär oder sekundär Primary
Gen-ID (Entrez) 79102
Zielspezies Human, Rat
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.