missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIZ1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
231.00 CHF - 549.00 CHF
Spezifikation
| Antigen | RIZ1 |
|---|---|
| Verdünnung | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Anwendungen | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
Beschreibung
RIZ1 Polyclonal antibody specifically detects RIZ1 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Spezifikation
| RIZ1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Chromatin Research | |
| PBS (pH 7.3), 50% glycerol | |
| 7799 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| EC 2.1.1.43, GATA-3 binding protein G3B, GATA-3-binding protein G3B, KMT8PR domain-containing protein 2, Lysine N-methyltransferase 8, MTB-ZFHUMHOXY1, PR domain containing 2, with ZNF domain, PR domain zinc finger protein 2, retinoblastoma protein-binding zinc finger protein, Retinoblastoma protein-interacting zinc finger protein, RIZ1, RIZ2, RIZMTE-binding protein, Zinc finger protein RIZ, zinc-finger DNA-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human PRDM2 (NP_036363.2). MNQNTTEPVAATETLAEVPEHVLRGLPEEVRLFPSAVDKTRIGVWATKPILKGKKFGPFVGDKKKRSQVKNNVYMWEVYYPNLGWMCIDATDPEKGNWLRYVNWACSGEEQNLFPLEINRAIYYKTLKPIAPGEELLVWYNGEDNPEIAAAIEEERASARSKRSSPKSRKGKKKSQENKNKGNKIQDIQLKTSEPDFTSANMRDSAEGPKEDEEKPSASALEQPATLQEVASQEVPPELA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts