Learn More
Brady™ Rigid Polypropylene Evacuation Panel Safety Signs
Marke: Brady™ 807240
Additional Details : Gewicht : 0.05000kg
Spezifikation
Polypropylene | |
Rigid Polypropylene | |
120°C | |
10 mm | |
Mini Pictograms Hazardous Label |
1 Ea. | |
210 mm | |
0.015 mm | |
Exit and Emergency Exit Up Arrow | |
White on Green |