Learn More
Rhodopsin Antibody (Rho 1D4), mFluor Violet 450 SE, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Rhodopsin Monoclonal antibody specifically detects Rhodopsin in Human, Mouse, Rat, Amphibian, Bovine, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune Assays
Spezifikation
Spezifikation
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 |
| Gen-ID (Entrez) | 6010 |
| Zielspezies | Mouse, Amphibian |