missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFX5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12137-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. Directly associates with HAX-1, through binding to its C-terminal region. May also be involved in the regulation of gene expression.Spezifikation
RFX5 | |
Polyclonal | |
Unconjugated | |
RFX5 | |
DNA binding protein RFX5, Regulatory factor X 5, RFX5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5993 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P48382 | |
RFX5 | |
RFX5 Fusion Protein Ag2783 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |