missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Respiratory Syncytial Virus Glycoprotein F Antibody (motavizumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
248.00 CHF - 1045.00 CHF
Spezifikation
| Antigen | Respiratory Syncytial Virus Glycoprotein F |
|---|---|
| Klon | motavizumab |
| Anwendungen | Flow Cytometry, ELISA, Functional Assay |
| Klassifikation | Monoclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30048048
|
Novus Biologicals™
NBP3-28280-1mg |
1 mg | |||||||
|
30046391
|
Novus Biologicals™
NBP3-28280-100ug |
100 μg |
407.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30044613
|
Novus Biologicals™
NBP3-28280-50ug |
50 μg |
248.00 CHF
50 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Respiratory Syncytial Virus Glycoprotein F Monoclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpezifikation
| Respiratory Syncytial Virus Glycoprotein F | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Virus | |
| RSV-F | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| motavizumab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts