missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zika Envelope F. Length Protein
A cDNA sequence encoding the Zika Envelope F. Length was constructed and used to recombinantly synthesize the protein.
227.00 CHF - 1735.00 CHF
Spezifikation
Name | Zika Envelope F. Length Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Zika |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15955369
|
enQuireBio™
QP13999-100UG |
100 μg |
227.00 CHF
100 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15975369
|
enQuireBio™
QP13999-500UG |
500 μg |
870.00 CHF
500 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15965369
|
enQuireBio™
QP13999-1MG |
1 mg |
1735.00 CHF
1 Milligramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
Zika Envelope F. Length Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Zika | |
His | |
Zika Envelope protein is >95% pure as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Zika Envelope protein solution contains PBS and 25mM arginine. |