missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zika Envelope F. Length Protein
A cDNA sequence encoding the Zika Envelope F. Length was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13999-500ug
Additional Details : Gewicht : 0.01000kg
Spécification
Zika Envelope F. Length Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Zika Envelope protein solution contains PBS and 25mM arginine. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Zika | |
His | |
Zika Envelope protein is >95% pure as determined by SDS-PAGE. |