missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant West Nile Virus WNV Pre-M Protein
A cDNA sequence encoding the WNV Pre-M was constructed and used to recombinantly synthesize the protein.
227.00 CHF - 1735.00 CHF
Spezifikation
Name | West Nile Virus WNV Pre-M Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | West Nile Virus |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15955259
|
enQuireBio™
QP13965-100UG |
100 μg |
227.00 CHF
100 Mikrogramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15975259
|
enQuireBio™
QP13965-500UG |
500 μg |
870.00 CHF
500 Mikrogramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15965259
|
enQuireBio™
QP13965-1MG |
1 mg |
1735.00 CHF
1 Milligramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
West Nile Virus WNV Pre-M Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
West Nile Virus | |
His | |
20mM phosphate buffer pH 7.5. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MVTLSNFQGKVMMTVNATDVTDVITIPTAAGKNLCIVRA MDVGYLCEDTITYECPVLAAGNDPEDIDCWCTKSSVYVRYGRCTKTRHSRRSRRSLTVQTHGESTLANKKGAWLDSTKATRYLVKTESWILRNPGYALE | |
Protein is >95% pure as determined by SDS-PAGE. |