missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Exfoliative toxin A Protein
A DNA sequence encoding the Staphylococcus aureus Exfoliative toxin A, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7304-ec-200ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
WP_001065781.1 | |
200 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |
S. aureus Exfoliative toxin A Protein | |
Research Use Only | |
eta | |
Recombinant Protein | |
E. coli | |
EVSAEEIKKH EEKWNKYYGV NAFNLPKELF SKVDEKDRQK YPYNTIGNVF VKGQTSATGV LIGKNTVLTN RHIAKFANGD PSKVSFRPSI NTDDNGNTET PYGEYEVKEI LQEPFGAGVD LALIRLKPDQ NGVSLGDKIS PAKIGTSNDL KDGDKLELIG YPFDHKVNQM HRSEIELTTL SRGLRYYGFT VPGNSGSGIF NSNGELVGIH SSKVSHLDRE HQINYGVGIG NYVKRIINEK NE | |
Greater than 80% as determined by SDS-PAGE. |