missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque IL 6 Protein
A cDNA sequence encoding the IL 6 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 6775.00 CHF
Spezifikation
Name | IL 6 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biologische Aktivität | Fully biologically active when compared to standard. The ED50 determined by a cell proliferation assay using IL-6-dependent murine 7TD1 cells is less than 0.1ng/ml, corresponding to a specific activity of > 1.0 M 10,000,000 IU/mg. |
Produkttyp | Recombinant Protein |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15984958
|
enQuireBio™
QP10719-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 25-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15964958
|
enQuireBio™
QP10719-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 25-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15974958
|
enQuireBio™
QP10719-1MG |
1 mg |
6775.00 CHF
1 Milligramm |
Verfügbar ab: 25-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
IL 6 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAPVLPGEDS KNVAAPHSQP LTSSERIDKH IRYILDGISA LRKETCNRSN MCESSKEALA ENNLNLPKMA EKDGCFQSGF NEDTCLVKII TGLLEFEVYL EYLQNRFESS EEQARAVQMS TKVLIQFLQK KAKNLDAITT PEPTTNASLL TKLQAQNQWL QDMTTHLILR SFKEFLQSNL RALRQM | |
Greater than 97.0% as determined by SDS-PAGE and HPLC analyses. |
Research Use Only | |
Fully biologically active when compared to standard. The ED50 determined by a cell proliferation assay using IL-6-dependent murine 7TD1 cells is less than 0.1ng/ml, corresponding to a specific activity of > 1.0 M 10,000,000 IU/mg. | |
Rhesus Macaque | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |