missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque IL 6 Protein
A cDNA sequence encoding the IL 6 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10719-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
IL 6 Protein | |
Research Use Only | |
Fully biologically active when compared to standard. The ED50 determined by a cell proliferation assay using IL-6-dependent murine 7TD1 cells is less than 0.1ng/ml, corresponding to a specific activity of > 1.0 M 10,000,000 IU/mg. | |
Rhesus Macaque | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAPVLPGEDS KNVAAPHSQP LTSSERIDKH IRYILDGISA LRKETCNRSN MCESSKEALA ENNLNLPKMA EKDGCFQSGF NEDTCLVKII TGLLEFEVYL EYLQNRFESS EEQARAVQMS TKVLIQFLQK KAKNLDAITT PEPTTNASLL TKLQAQNQWL QDMTTHLILR SFKEFLQSNL RALRQM | |
Greater than 97.0% as determined by SDS-PAGE and HPLC analyses. |