missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat GCSF Protein
A cDNA sequence encoding the GCSF was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10625-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
GCSF Protein | |
Research Use Only | |
The ED50 determined by a cell proliferation assay using murine NFS-60 cells is less than 0.05ng/ml, corresponding to a specific activity of > 2.0M 107 IU/mg. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in 5mM Sodium Citrate, pH 4.0. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
KKIPLLTVSS LPPSLPLPRS FLLKSLEQVR KIQARNTELL EQLCATYKLC HPEELVLFGH SLGIPKASLS SCSSQALQQT KCLSQLHSGL FLYQGLLQAL AGISSELAPT LDMLHLDVDN FATTIWQQME SLGVAPTVQP TQSTMPIFTS AFQRRAGGVL VTSYLQSFLE TAHHALHHLP RPAQKHFPES LFISI | |
Greater than 97.0% as determined by:(a)Analysis by RP-HPLC. (b)Analysis by SDS-PAGE. |