missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL28 Protein
A DNA sequence encoding the Rat CCL28, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10300-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_446152.1 | |
CCL28 Protein | |
Research Use Only | |
Ccl28 | |
The biologically active determined by a chemotaxis bioassay using human lymphocytes is in a concentration range of 5.0-50 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
114492 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SEAILPIASS CCTEVSHHIP RRLLERVNSC SIQRADGDCD LAAVILHVKR RRICVSPHNP TLKRWMSASE MKNGKENLCP RKKQDSGKDR KGHTPRKHGK HGTRRIHGTH DHEAPR | |
Greater than 96% as determined by SDS-PAGE and HPLC. |